Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EBI3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA523297

Catalog No. PIPA523297


Add to cart

Description

Description

EBI3 is a 228 amino acid containing pro- and anti-inflammatory cytokine belonging to the type I cytokine receptor family and contains two fibronectin type-III domain. It forms a heterodimer with IL-27 and is also a component of the IL-12 heterodimer. A protein involved in regulated cell-mediated immune responses, EBI3 has an important role in regulating T helper cell development, suppressing T-cell proliferation, stimulating cytotoxic T-cell activity, inducing isotype switching in B-cells, and has diverse effects on innate immune cells. Target cells of EBI3 include CD4 T helper cells, type 1 effector cells (TH1), type 2 effector cells (TH2) and IL-17 producing helper T-cells (TH17). It drives rapid clonal expansion of naive but not memory CD4 T-cells. EBI3 is important in anti-tumor as well as anti-angiogenic activity with activation and production of anti-angiogenic chemokines. It also strongly synergizes with IL-12 to trigger interferon-gamma/IFN-gamma production of naive CD4 T-cells.
Specifications

Specifications

EBI3 
Polyclonal
Unconjugated
EBI3
Liquid
EBI3
Synthetic peptide (DLTDYGKPSDWSLPGQVEc (aa205-222)) of EBI3  protein.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Immunohistochemistry (Paraffin), Western Blot
1 mg/mL
PBS with 0.05% sodium azide
Q14213, O35228
IL-27B, IL27B, EBV-induced gene 3 protein, Epstein-Barr virus induced gene 3, IL-27 subunit beta, IL27 subunit, IL35 subunit, cytokine receptor, epstein-Barr virus-induced gene 3 protein, interleukin-27 subunit beta
Rabbit
IgG
100 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
10148, 50498, 680609
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit