Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EBI3 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA523228

Catalog No. PIPA523228


Add to cart

Description

Description

EBI3 is a novel hematopoietin receptor family member related to the p40 subunit of interleukin-12 and to the ciliary neurotrophic factor receptor. It ft.s expression is induced in B lymphocytes by Epstein-Barr virus (EBV) infection. Human EBI3 gene maps to chromosome 19p13.3, near genes encoding the erythropoietin receptor and the cytokine receptor-associated kinase, Tyk2. The protein is expressed in vivo by scattered cells in interfollicular zones of tonsil tissue, by cells associated with sinusoids in perifollicular areas of spleen tissue, and at very high levels by placental syncytiotrophoblasts. EBI3 expression in vitro is induced in EBV-negative cell lines by expression of the EBV latent infection membrane protein-1 and in peripheral blood mononuclear cells by pokeweed mitogen stimulation. The protein plays a crucial role in regulating cell-mediated immune responses.
Specifications

Specifications

EBI3
Polyclonal
Unconjugated
EBI3
Liquid
EBI3
Synthetic peptide (73 RGHSWPCLQQTPTSTSC 89) of EBI3 protein.
Protein G
RUO
Antibody
Polyclonal
Human, Mouse
Western Blot
0.5 mg/mL
PBS with 0.05% BSA and 0.05% sodium azide
Q14213, O35228
IL-27B, IL27B, EBV-induced gene 3 protein, Epstein-Barr virus induced gene 3, IL-27 subunit beta, IL27 subunit, IL35 subunit, cytokine receptor, epstein-Barr virus-induced gene 3 protein, interleukin-27 subunit beta
Rabbit
IgG
100 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
10148, 50498
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit