Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EBP50 Mouse anti-Human, Unlabeled, Clone: 6, BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 611160

 View more versions of this product

Catalog No. BDB611160


Add to cart

Description

Description

Integral plasma membrane proteins are stabilized by linkages to the cortical actin cytoskeleton, which structurally supports the membrane and contributes to processes such as endocytosis, exocytosis, and transmembrane signaling. The ERM (ezrin-radixin-moesin) family of proteins provides these structural linkages. The ERM proteins contain a 300-residue N-terminal domain, a 170-residue α-helical region, and a C-terminal 100-residue domain that contains F-actin binding sites. The N-terminal domain interacts with the cytoplasmic domain of CD44, the regulatory subunit of PKA, and the PDZ domain containing ERM-binding phosphoprotein (EBP-50). In polarized epithelial cells, EBP-50 links ezrin and the cytoplasmic regions of transmembrane proteins such as the cystic fibrosis transmembrane conductance regulator (CFTR) and the β2-adrenergic receptor. EBP50 contains two PDZ domains followed by a C-terminal tail. It colocalizes with ezrin in apical microvilli and is also thought to interact with the Na+-H+ exchanger NHE3 to confer cAMP-mediated inhibition of Na+-H+ exchange. Thus, EBP-50 mediates membrane attachment to the cytoskeleton and may also function in the regulation of ion exchange.

Host Species: Mouse
Clone: 6
Isotype: IgG1
Species Reactivity: Mouse
Immunogen: Human EBP50 aa. 128-249
Formula Weight [Chemical]: 50-53kDa

Immunofluorescence, Western Blotting

Specifications

Specifications

EBP50
6
Unconjugated
Affinity Purified
Human EBP50 aa. 128-249
50μg
Cell Biology
Primary
Human
Western Blot
250μg/mL
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Mouse
IgG1
RUO
Store undiluted at -20°C.
Monoclonal
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only.