Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EBPL Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA525639

Catalog No. PIPA525639

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

EBPL does not possess sterol isomerase activity and does not bind sigma ligands.
Specifications

Specifications

EBPL
Polyclonal
PBS with 0.09% sodium azide
Q9BY08
EBRP, emopamil binding related protein, delta8-delta7 sterol isomerase related protein, emopamil-binding protein-like, emopamil-binding-related protein
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
84650
Flow Cytometry, Immunohistochemistry, Western Blot
Unconjugated
EBPL
Liquid
EBPL
KLH conjugated synthetic peptide between 177-206 amino acids from the C-terminal region of human EBPL
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit