Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ECE1 Rat anti-Human, Clone: 303913, Invitrogen™

Rat Monoclonal Antibody

Manufacturer:  Invitrogen MA524016

Catalog No. PIMA524016


Add to cart

Description

Description

Reconstitute at 0.5 mg/mL in sterile PBS.

ECE-1 is involved in proteolytic processing of endothelin precursors to biologically active peptides.
Specifications

Specifications

ECE1
Monoclonal
0.5 mg/mL
PBS with 5% trehalose and No Preservative
P42892
ECE, ECE-1, endothelin-converting enzyme 1
Rat
IgG1
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
1889
Flow Cytometry
303913
Unconjugated
ECE1
Lyophilized
ECE1
Mouse myeloma cell line NS0-derived recombit human ECE-1 Gln90-Trp770
Protein A/G
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit