Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ECHDC1 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA526663
Description
This antibody is predicted to react with rat based on sequence homology.
ECHDC1 belongs to the enoyl-CoA hydratase/isomerase family. There are two named isoforms.Specifications
ECHDC1 | |
Polyclonal | |
ECHDC1 | |
dJ351K20.2, enoyl Coenzyme A hydratase domain containing 1, enoyl-CoA hydratase domain-containing protein 1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
52665, 55862 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
Unconjugated | |
Q9NTX5, Q9D9V3 | |
ECHDC1 | |
KLH conjugated synthetic peptide between 254-283 amino acids from the C-terminal region of human ECHDC1 | |
400 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction