Learn More
Eck/EphA2 Mouse anti-Human, Rat, Mouse, Bovine, Canine, Clone: D7, MilliporeSigma™
Mouse Monoclonal Antibody
Manufacturer:  MilliporeSigma 05480
Catalog No. 05-480-MI
Add to cart
Description
Description
This Anti-Eck/EphA2 Antibody, clone D7 is validated for use in immunocytochemistry, immunoprecipitation, EA, western blotting for the detection of Eck/EphA2.
Specifications
Specifications
Eck/EphA2
D7
Purified mouse monoclonal IgG1 in buffer containing 0.2MTris-glycine, pH 7.4, 0.15MNaCl, with 0.05% sodium azide and 30% glycerol.
Purified
Murine
IgG1
200μg
Primary
Bovine, Canine, Human, Murine, Rat
ELISA, Immunocytochemistry, Immunoprecipitation, Western Blot
Unconjugated
NM_004431
EPHA2, EphA2, ECK
Native protein isolated by purification of phosphotyrosine-containing proteins. Clone D7.
Protein G purfied
−20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.
Monoclonal