Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-ECT2, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP128729

Catalog No. NBP128729


Add to cart

Description

Description

ECT2 Polyclonal antibody specifically detects ECT2 in Human samples. It is validated for Western Blot, Immunoprecipitation (Negative).
Specifications

Specifications

ECT2
1.0 mg/mL
Western Blot 1:2000-1:10000, Immunoprecipitation
Q9H8V3
ARHGEF31, epithelial cell transforming sequence 2 oncogene, Epithelial cell-transforming sequence 2 oncogene, FLJ10461, MGC138291, protein ECT2
Rabbit
IgG
Immunogen affinity purified
RUO
Primary
1894
Immunoprecipitation, Western Blot
Unconjugated
Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide
Affinity Purified
ECT2
The immunogen recognized by this antibody maps to a region between residue 833 and 883 of human epithelial cell transforming sequence 2 using the numbering given in entry NP_060568.3 (GeneID 1894).
104 kDa
100 ul
Store at 4C. Do not freeze.
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only