Learn More
ectodysplasin A2 receptor, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a full-length human EDA2R protein.
Manufacturer: Abnova Corporation H00060401B01
Description
EDA-A1 and EDA-A2 are two isoforms of ectodysplasin that are encoded by the anhidrotic ectodermal dysplasia (EDA) gene. Mutations in EDA give rise to a clinical syndrome characterized by loss of hair, sweat glands, and teeth. The protein encoded by this gene specifically binds to EDA-A2 isoform. This protein is a type III transmembrane protein of the TNFR (tumor necrosis factor receptor) superfamily, and contains 3 cysteine-rich repeats and a single transmembrane domain but lacks an N-terminal signal peptide. Multiple alternatively spliced transcript variants have been found for this gene, but some variants lack sufficient support. [provided by RefSeq
Sequence: MDCQENEYWDQWGRCVTCQRCGPGQELSKDCGYGEGGDAYCTACPPRRYKSSWGHHRCQSCITCAVINRVQKVNCTATSNAVCGDCLPRFYRKTRIGGLQDQECIPCTKQTPTSEVQCAFQLSLVEADAPTVPPQEATLVALVSSLLVVFTLAFLGLFFLYCKQFFNRHCQRGGLLQFEADKTAKEESLFPVPPSKETSAESQVSENIFQTQPLNPILEDDCSSTSGFPTQESFTMASCTSESHSHWVHSPIECTELDLQKFSSSASYTGAETLGGNTVESTGDRLELNVPFEVPSPSpecifications
ectodysplasin A2 receptor | |
Polyclonal | |
Mouse polyclonal antibody raised against a full-length human EDA2R protein. | |
EDA2R | |
EDA-A2R/EDAA2R/TNFRSF27/XEDAR | |
Mouse | |
50 μL | |
Membrane Receptors | |
Primary | |
Human |
Flow Cytometry, Western Blot | |
Unconjugated | |
No additive | |
NM_021783 | |
EDA2R | |
EDA2R (NP_068555, 1 a.a. ∼ 297 a.a) full-length human protein. | |
RUO | |
Yes | |
60401 | |
Serum |
Safety and Handling
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.