Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EDG3 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA112703

Catalog No. 01-674-841


Add to cart

Description

Description

A suggested positive control for this product is RH7777 cells transfected with EDG3 protein.

Endothelial cell differentation gene-3 (EDG3) belongs to a family of G-protein couple receptors whose ligands are lysophospholipids. The ligand for EDG3 is sphingosine-1-phosphate. There are 8 known members of the EDG receptor family. They have been implicated in mediating growth related effects such as induction of cellular proliferation, alterations in differentiation and survival suppression of apoptosis. They also evoke cellular effector functions that are dependent on cytoskeletal responses such as contraction, secretion, adhesion and chemotaxis. EDG receptors are developmentally regulated and differ in tissue distribution. The couple to multiple types of G proteins to signal through ras and MAP kinase, rho, phospholipase C and several protein tyrosine kinases. EDG3 is expressed in cardiovascular, leukocyte-containing and other tissues.
Specifications

Specifications

EDG3
Polyclonal
Unconjugated
S1PR3
Liquid
S1PR3
Synthetic peptide corresponding to the N terminal of EDG3 (S1P3).
Ammonium sulfate precipitation
RUO
Antibody
Polyclonal
Human
Western Blot
1 mg/mL
PBS with 0.08% sodium azide
Q99500
S1P3
Rabbit
IgG
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
1903
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit