Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EDG6 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA112706

Catalog No. 01-674-844


Add to cart

Description

Description

A suggested positive control for this product is COS cells transfected with EDG6.

EDG6 is the receptor for lysosphingolipid sphingosine 1 phosphate (S1P). S1P is a bioactive lysophospholipid that elicits diverse physiological effect on most types of cells and tissues. EDG6 may be involved in cell migration processes that are specific for lymphocytes.
Specifications

Specifications

EDG6
Polyclonal
PBS with 0.08% sodium azide
O95977
S1P4
Rabbit
IgG
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
8698
Western Blot
Unconjugated
S1PR4
Liquid
S1PR4
Synthetic peptide corresponding to the N terminal of EDG6 (S1P4) receptor.
Ammonium sulfate precipitation
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit