Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-EDIL3/DEL1, FITC, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP128632F

Catalog No. NBP128632F


Add to cart

Description

Description

EDIL3/DEL1 Polyclonal antibody specifically detects EDIL3/DEL1 in Human, Mouse samples. It is validated for Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

EDIL3/DEL1
FITC
PBS with 0.05% Sodium Azide
Affinity Purified
EDIL3
Synthetic peptide made to an internal portion of human Del-1 (within residues 200-350). [Swiss-Prot# O43854]
53 kDa
0.1 ml
Angiogenesis, Cytoskeleton Markers, Endothelial Cell Markers, Extracellular Matrix, Signal Transduction
Primary
10085
Immunocytochemistry, Immunofluorescence
Immunocytochemistry/Immunofluorescence
O43854
DEL1developmental endothelial locus-1, Developmentally-regulated endothelial cell locus 1 protein, EGF-like repeat and discoidin I-like domain-containing protein 3, EGF-like repeats and discoidin I-like domains 3, Integrin-binding protein DEL1, MGC26287
Rabbit
IgG
Immunogen affinity purified
RUO
Store at 4C in the dark.
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only