Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-EDNRB/Endothelin R Type B, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP131108

Catalog No. NBP131108


Add to cart

Description

Description

EDNRB/Endothelin R Type B Polyclonal antibody specifically detects EDNRB/Endothelin R Type B in Human, Rat, Bovine samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

EDNRB/Endothelin R Type B
0.52mg/mL
Western Blot 1:500-1:3000, Immunohistochemistry 10 - 1:500, Immunohistochemistry-Paraffin 1:100-1:1000
P24530
ABCDS, endothelin B receptor, Endothelin receptor non-selective type, endothelin receptor type B, ETB, ET-B, ETBR, ET-BR, ETRB, HSCR, HSCR2, WS4A
Rabbit
IgG
Antigen Affinity-purified
RUO
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Polyclonal
Not yet tested in any other species.
Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol with 0.01% Thimerosal
Affinity Purified
EDNRB
Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human Endothelin B Receptor. The exact sequence is proprietary.
50 kDa
0.1 ml
GPCR, Neuroscience, Signal Transduction
Primary
1910
Bovine, Human, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only