Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EEF1D Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA527666

Catalog No. PIPA527666


Add to cart

Description

Description

PA5-27666 targets EEF1D in IF and WB applications and shows reactivity with Human and mouse samples. The PA5-27666 immunogen is recombit fragment corresponding to a region within amino acids 1 and 281 of Human EEF1D. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

This gene encodes a subunit of the elongation factor-1 complex, which is responsible for the enzymatic delivery of aminoacyl tRNAs to the ribosome. This subunit functions as guanine nucleotide exchange factor. It is reported that this subunit interacts with HIV-1 Tat, and thus it represses the translation of host-cell, but not HIV-1, mRNAs. Several alternatively spliced transcript variants encoding multiple isoforms have been found for this gene.
Specifications

Specifications

EEF1D
Polyclonal
Unconjugated
EEF1D
Liquid
EEF1D
Recombit fragment corresponding to a region within amino acids 1 and 281 of Human EEF1D
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Immunocytochemistry, Immunofluorescence, Western Blot
0.91 mg/mL
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
P29692, P57776
EF-1D, EF1D, FP1047, EF-1-delta, antigen NY-CO-4, elongation factor 1-delta, guanine nucleotide exchange protein
Rabbit
IgG
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
1936, 66656
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit