Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EEFSEC Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA526085

Catalog No. PIPA526085

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

EEFSEC is a translation factor necessary for the incorporation of selenocysteine into proteins. It probably replaces EF-Tu for the insertion of selenocysteine directed by the UGA codon. SelB binds GTP and GDP.
Specifications

Specifications

EEFSEC
Polyclonal
PBS with 0.09% sodium azide
P57772, Q9JHW4
EFSEC, SELB, elongation factor for selenoprotein translation, elongation factor sec, selenocysteine-specific elongation factor
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
60678, 65967
Immunohistochemistry, Western Blot
Unconjugated
EEFSEC
Liquid
EEFSEC
KLH conjugated synthetic peptide between 541-568 amino acids from the C-terminal region of human EEFSEC
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit