Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fibulin 3 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA526104

Catalog No. PIPA526104


Add to cart

Description

Description

This antibody is predicted to react with non-human primate and rat based on sequence homology.

EFEMP1 encodes a member of the fibulin family of extracellular matrix glycoproteins. Like all members of this family, the encoded protein contains tandemly repeated epidermal growth factor-like repeats followed by a C-terminus fibulin-type domain. This gene is upregulated in maligt gliomas and may play a role in the aggressive nature of these tumors.
Specifications

Specifications

Fibulin 3
Polyclonal
PBS with 0.09% sodium azide
Q12805, Q8BPB5
DHRD, DRAD, FBLN3, FBNL, MLVT, MTLV, S1-5, EGF-containing fibulin-like extracellular matrix protein 1, FIBL-3, extracellular protein S1-5, fibulin-3
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
216616, 2202
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
EFEMP1
Liquid
EFEMP1
KLH conjugated synthetic peptide between 119-148 amino acids from the N-terminal region of human EFEMP1
Antigen Affinity Chromatography, Protein A
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit