Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EFEMP2 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA526277

Catalog No. PIPA526277

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

This antibody is predicted to react with hamster based on sequence homology.

EFEMP2 have been found to contain variations of the epidermal growth factor (EGF) domain and have been implicated in functions as diverse as blood coagulation, activation of complement and determination of cell fate during development. The protein contains four EGF2 domains and six calcium-binding EGF2 domains. This protein is necessary for elastic fiber formation and connective tissue development.
Specifications

Specifications

EFEMP2
Polyclonal
PBS with 0.09% sodium azide
O95967, Q9WVJ9
ARCL1B, FBLN4, MBP1, UPH1, EGF-containing fibulin-like extracellular matrix protein 2, FIBL-4, fibulin 4, fibulin-4, mutant p53 binding protein 1
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
30008, 58859
Flow Cytometry, Western Blot
Unconjugated
EFEMP2
Liquid
EFEMP2
KLH conjugated synthetic peptide between 386-414 amino acids from the C-terminal region of human EFEMP2
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit