Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EFHC2 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA526761

Catalog No. PIPA526761

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

EFHC2
Polyclonal
PBS with 0.09% sodium azide
Q5JST6
dJ1158H2.1, EF-hand domain-containing family member C2
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
80258
Flow Cytometry, Western Blot
Unconjugated
EFHC2
Liquid
EFHC2
KLH conjugated synthetic peptide between 93-122 amino acids from the N-terminal region of human EFHC2
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit