Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EFTUD1 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA523877

Catalog No. PIPA523877

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

EFTUD1
Polyclonal
PBS with 0.09% sodium azide
Q7Z2Z2, Q8C0D5
FAM42A, HsT19294, RIA1, elongation factor Tu GTP-binding domain-containing protein 1, elongation factor-like 1, ribosome assembly 1 homolog
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
101592, 79631
Flow Cytometry, Immunohistochemistry, Western Blot
Unconjugated
EFL1
Liquid
EFL1
KLH conjugated synthetic peptide between 1062-1090 amino acids from the C-terminal region of human EFTUD1
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit