Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EFTUD2 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA526296

Catalog No. PIPA526296

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

This antibody is predicted to react with bovine, chicken and mouse based on sequence homology.

EFTUD2 is component of the U5 snRNP complex required for pre-mRNA splicing.
Specifications

Specifications

EFTUD2
Polyclonal
PBS with 0.09% sodium azide
Q15029
Snrp116, Snu114, U5-116KD, 116 kDa U5 small nuclear ribonucleoprotein component, SNU114 homolog, U5 snRNP specific protein, 116 kD, U5 snRNP-specific protein, 116 kD, U5 snRNP-specific protein, 116 kDa, U5-116 kDa, elongation factor Tu GTP-binding domain-containing protein 2, hSNU114
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
9343
Immunohistochemistry, Western Blot
Unconjugated
EFTUD2
Liquid
EFTUD2
KLH conjugated synthetic peptide between 257-284 amino acids from the central region of human EFTUD2
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit