Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eg5 Mouse anti-Human, Unlabeled, Clone: 20, BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 611186

 View more versions of this product

Catalog No. BDB611186


Add to cart

Description

Description

The mitotic spindle apparatus equally distributes duplicated chromosomes to daughter cells. This process is mediated by the dynamic instability of the spindle microtubules and the forces generated by the action of cytoplasmic dynein and kinesin-related motor proteins (KRPs), such as CHO1/MKLP1. These motors hydrolyze ATP as they move progressively along the microtubules. They organize the microtubules into bipolar spindles and may also serve as bridges between microtubules and chromosome kinetochores or centrosomes. Eg5, identified in Xenopus laevis, is a plus-end directed KRP, associated with the mitotic spindle. Vertebrate Eg5 is a member of the KRP subfamily bimC. These family members share a high degree of sequence homology (>80% within the N-terminal motor domain) and may execute similiar functions. During mitosis, Eg5 is specifically phosphorylated at Thr-297, an evolutionarily conserved cdc2 phosphorylation site, by p34[cdc2]/cyclin B. Inhibition of phosphorylation blocks the interaction of Eg5 with centrosomes. Thus, Eg5 is a mitotic motor protein that regulates spindle formation in a phosphorylation-dependent manner.

Host Species: Mouse
Clone: 20
Isotype: IgG1
Species Reactivity: Human
Immunogen: Human Eg5 aa. 324-532
Formula Weight [Chemical]: 120kDa

Immunofluorescence, Western Blotting

Specifications

Specifications

Eg5
20
Unconjugated
Affinity Purified
Human Eg5 aa. 324-532
50μg
Cell Biology
Primary
Human
Western Blot
250μg/mL
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Murine
IgG1
RUO
Store undiluted at -20°C.
Monoclonal
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only.