Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-EGF, Polyclonal, Abcam™ Non-distribution product as customer accommodation.

Manufacturer:  Abcam AB9695100UG

Catalog No. NC0179582


Add to cart

Description

Description

Host Species: Rabbit
Isotype: IgG
Cross Reactivity: Human
Immunogen: Highly pure (>98%) recombinant hEGF (human Epidermal Growth Factor)

Applications:

ELISA, Immunohistochemistry (Paraffin), Western Blotting

Specifications

Specifications

EGF
Unconjugated
Rabbit
IgG
Primary
Human
0.5mg/mL
Affinity Purified
Highly pure (>98%) recombinant hEGF (human Epidermal Growth Factor)
100μg
Polyclonal
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit