Learn More
EGF Receptor Mouse anti-Human, Brilliant Violet 510, Clone: EGFR.1, BD
Mouse Monoclonal Antibody
Manufacturer:  BD Biosciences 563344
Catalog No. BDB563344
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
The EGFR.1 monoclonal antibody specifically binds to the epidermal growth factor receptor (EGF-R). The EGF-R is a transmembrane glycoprotein ofapproximately 170 kDa that is expressed on most cells. It consists of a glycosylated extracellular domain which binds to epidermal growth factor (EGF), a transmembrane domain, and an intracellular domain with tyrosine-kinase activity essential for signal transduction. The EGF-R plays an important role in the growth and differentiation of many cellular types. Transforming growth factor α (TGFα), vaccinia virus growth factor, and related growth factors can also bind to and signal through the EGF-R.
Specifications
Specifications
EGF Receptor
EGFR.1
EGFR; ERBB; ERBB1; HER1; mENA; PIG61
Affinity Purified
IgG2b, κ
RUO
Primary
Human
Flow Cytometry
Brilliant Violet 510
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Murine
50 Tests
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Monoclonal