Learn More
EGFR (CT) Mouse anti-Human, Mouse, Clone: 13G8, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier:  MilliporeSigma 04290
Catalog No. 04-290-MI
Add to Cart
Description
Description
Anti-EGFR (C-terminus) Antibody, clone 13G8 detects level of EGFR (C-terminus) and has been published and validated for use in ELISA, IC and western blotting.
Specifications
Specifications
EGFR (CT)
Monoclonal
Unconjugated
NM_005228.3; NM_201282.1; NM_201283.1; NM_201284.1
Mouse
Subsequent thiophilic adsorption and size exclusion chromatography
Primary
−20°C for 24 months.
IgG1
ELISA, Immunocytochemistry, Western Blot
13G8
100μg of purified mouse monoclonal IgG lyophilized in 2X PBS containing 0.09% sodium azide, PEG, and sucrose
EGFR, ERBB1, ERBB, mENA
KLH-conjugated synthetic peptide encompassing the surrounding amino acids of the C terminus in human EGFR
100 μg
Human, Mouse
Purified