Learn More
EGFR (non-phospho-Tyr1173) Mouse anti-Human, Mouse, Clone: 20G3, MilliporeSigma™
Mouse Monoclonal Antibody
Manufacturer:  MilliporeSigma 05484
Catalog No. 05-484-MI
Add to cart
Description
Description
Detect EGFR (non-phospho-Tyr1173) using this Anti-EGFR (non-phospho-Tyr1173) Antibody, clone 20G3 validated for use in immunoprecipitation and western blotting.
Specifications
Specifications
EGFR (non-phospho-Tyr1173)
20G3
100μg of lyophilized thiophilic and size exclusion chromatography purified mouse IgG1 from 1mL 2x PBS / 0.09 % Na-azide / PEG and Sucrose. Reconstitute with 100μL H2O (15 min, RT) for a final concentration of 1mg/mL.
Purified
Murine
κ
100μg
Primary
Human, Murine
Immunoprecipitation, Western Blot
Unconjugated
NM_005228
EGFR, ERBB1, ERBB, mENA
peptide containing the unphosphorylated Tyr1173 motif (NAE[Y]LR) of the human EGF receptor
Thiophilic adsoption and size exclusion chromatography
−20°C for 24 months.
Monoclonal