Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EGLN1/PHD2 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 33 publications
Supplier: Novus Biologicals NB1002219
Description
EGLN1/PHD2 Polyclonal specifically detects EGLN1/PHD2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Knockout Validated, Knockdown Validated.Specifications
EGLN1/PHD2 | |
Polyclonal | |
Unconjugated | |
PBS with 0.02% Sodium Azide | |
EGLN1 | |
A synthetic peptide made to an internal portion of mouse PHD2/HIF Prolyl Hydroxylase 2 (between residues 300-400). [Uniprot: Q91YE3] | |
Affinity Purified | |
RUO | |
Primary | |
Human and Mouse. Predicted to react with rat based on 100% sequence homology. | |
Store at 4C. Do not freeze. |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunoassay, KnockDown | |
1 mg/mL | |
Western Blot 2 ug/mL, Immunohistochemistry 2.5 - 5.0 ug/mL, Immunocytochemistry/Immunofluorescence 1:50 - 1:500, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Paraffin 2.5 - 5.0 ug/mL, Knockout Validated reported in scientific literature (PMID 24695462), KnockDown Validated | |
C1orf12HPH2, EC 1.14.11, EC 1.14.11.-, ECYT3, egl nine homolog 1, egl nine homolog 1 (C. elegans), egl nine-like protein 1, HIF prolyl hydroxylase 2, HIF-PH2, HIFPH2 EGL nine (C.elegans) homolog 1, HIF-prolyl hydroxylase 2, HPH-2, Hypoxia-inducible factor prolyl hydroxylase 2, PHD2SM20, Prolyl hydroxylase domain-containing protein 2, SM-20, zinc finger MYND domain-containing protein 6, ZMYND6DKFZp761F179 | |
Rabbit | |
43 kDa | |
0.1 mL | |
Cancer, Chromatin Research, Hypoxia | |
54583 | |
Human, Mouse, Rat | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction