Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-EGLN1/PHD2, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 33 publications

Manufacturer:  Novus Biologicals NB1002219SS

 View more versions of this product

Catalog No. NB1002219SS


Add to cart

Description

Description

EGLN1/PHD2 Polyclonal specifically detects EGLN1/PHD2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Knockout Validated, Knockdown Validated.
Specifications

Specifications

EGLN1/PHD2
Polyclonal
Unconjugated
PBS with 0.02% Sodium Azide
EGLN1
A synthetic peptide made to an internal portion of mouse PHD2/HIF Prolyl Hydroxylase 2 (between residues 300-400). [Uniprot: Q91YE3]
Affinity Purified
RUO
Primary
Human and Mouse. Predicted to react with rat based on 100% sequence homology.
Store at 4C. Do not freeze.
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunoassay, KnockDown
1 mg/mL
Western Blot 2 ug/mL, Immunohistochemistry 2.5 - 5.0 ug/mL, Immunocytochemistry/Immunofluorescence 1:50 - 1:500, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Paraffin 2.5 - 5.0 ug/ml, Knockout Validated, Knockdown Validated
C1orf12HPH2, EC 1.14.11, EC 1.14.11.-, ECYT3, egl nine homolog 1, egl nine homolog 1 (C. elegans), egl nine-like protein 1, HIF prolyl hydroxylase 2, HIF-PH2, HIFPH2 EGL nine (C.elegans) homolog 1, HIF-prolyl hydroxylase 2, HPH-2, Hypoxia-inducible factor prolyl hydroxylase 2, PHD2SM20, Prolyl hydroxylase domain-containing protein 2, SM-20, zinc finger MYND domain-containing protein 6, ZMYND6DKFZp761F179
Rabbit
43 kDa
0.025 mL
Cancer, Chromatin Research, Hypoxia
54583
Human, Mouse, Rat
IgG
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only