Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-EGR2, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$155.80 - $361.50

Specifications

Host Species Rabbit
Isotype IgG
Immunogen A synthetic peptide made to a portion of human EGR2 (within residues 200-300). [Swiss-Prot# P11161]
Antigen EGR2
Monoclonal or Polyclonal Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB11059723T
View Documents
Novus Biologicals
NB11059723SS
0.025 ml
Each for $155.80
Add to cart
 
NB11059723
View Documents
Novus Biologicals
NB11059723
0.1 ml
Each for $361.50
Add to cart
 
Description

Description

EGR2 Polyclonal antibody specifically detects EGR2 in Human, Mouse, Porcine samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Immunofluorescence.
Specifications

Specifications

Rabbit
A synthetic peptide made to a portion of human EGR2 (within residues 200-300). [Swiss-Prot# P11161]
Polyclonal
Primary
Western Blot, Immunocytochemistry, Immunofluorescence
P11161
PBS with 0.05% Sodium Azide
IgG
EGR2
Affinity Purified
RUO
Human and porcine. Immunogen sequence has 86% homology to mouse, rat, chicken and Xenopus proteins.
1959
1.0 mg/mL
SDS
Product Certifications

For Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit