Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EHMT2 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA525440

Catalog No. PIPA525440

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

A cluster of genes, BAT1-BAT5, has been localized in the vicinity of the genes for TNF alpha and TNF beta. This gene is found near this cluster; it was mapped near the gene for C2 within a 120-kb region that included a HSP70 gene pair. These genes are all within the human major histocompatibility complex class III region. This gene was thought to be two different genes, NG36 and G9a, adjacent to each other but a recent publication shows that there is only a single gene. The protein encoded by this gene is thought to be involved in intracellular protein-protein interaction.
Specifications

Specifications

EHMT2
Polyclonal
PBS with 0.09% sodium azide
Q96KQ7
BAT8, C6orf30, G9A, GAT8, KMT1C, NG36, G9A histone methyltransferase, H3-K9-HMTase 3, HLA-B associated transcript 8, HLA-B-associated transcript 8, ankyrin repeat-containing protein, histone H3-K9 methyltransferase 3, histone-lysine N-methyltransferase EHMT2, histone-lysine N-methyltransferase, H3 ly
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
10919
Flow Cytometry, Western Blot
Unconjugated
EHMT2
Liquid
EHMT2
KLH conjugated synthetic peptide between 159-186 amino acids from the N-terminal region of human EHMT2
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit