Learn More
eIF-4γ Mouse, Unlabeled, Clone: 40, BD
Mouse Monoclonal Antibody
Supplier: BD Biosciences 610536
Description
eIF-4 proteins are required for mRNA recognition and acceleration of translation during protein synthesis. This group of proteins consists of the RNA helicase, eIF-4A; the RNA-binding protein, eIF-4B; and the cap binding proteins, eIF-4E and eIF-4γ. eIF-4γ, also referred to as p220, is a 1396 amino acid polypeptide that migrates as a 220kDa. eIF-4γ has several potential glycosylation and phosphorylation sites that could account for its anomalous electrophoretic mobility. In addition, eIF-4γ is rapidly degraded in picornavirus-infected cells due to its content of multiple PEST sequences.
Specifications
eIF-4γ | |
Monoclonal | |
250μg/mL | |
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
Human eIF-4γ aa. 1217-1386 | |
50 μg | |
Cell Biology | |
Canine, Human | |
IgG1 |
Western Blot | |
40 | |
Unconjugated | |
Mouse | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at -20°C. |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.