Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

eIF-4E Mouse, FITC, Clone: 87, BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 611624

Catalog No. BDB611624


Add to cart

Specifications

Specifications

eIF-4E
87
FITC
Affinity Purified
Rabbit eIF-4E aa. 1-217
50μg
Cell Biology
Primary
Canine, Chicken, Frog, Human, Murine, Rat
In vitro Assay
250μg/mL
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Murine
IgG1
RUO
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Monoclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit