Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

eIF2b beta, Polyclonal, Invitrogen™

Chicken Polyclonal Antibody

Manufacturer:  Invitrogen PA127795

Catalog No. PIPA127795

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Catalyzes the exchange of eukaryotic initiation factor 2-bound GDP for GTP. Eukaryotic initiation factor-2B (EIF2B) is a GTP exchange protein essential for protein synthesis. It consists of alpha (EIF2B1; MIM 606686). beta (EIF2B2). gamma (EIF2B3; MIM 606

PA1-27795 detects EIF2B2 from human and mouse samples.PA1-27795 may be successfully used in ELISA and Western blot applications. By Western blot, this antibody has only been tested on fusion protein.The immunogen is a synthetic peptide corresponding to human Translation initiation factor eIF-2B subunit beta.
Specifications

Specifications

EIF2B2
Polyclonal
PBS with no preservative
P49770, Q99LD9
EIF-2Bbeta, EIF2B
Chicken
IgY
50μg
Store at -20°C
Primary
8892
ELISA, Western Blot
Unconjugated
EIF2B2
Purified
EIF2B2
Synthetic peptide corresponding to human Translation initiation factor eIF-2B subunit beta.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Murine
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit