Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

eIF3b Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA523278

Catalog No. PIPA523278


Add to cart

Description

Description

eIF3B is an acidic protein belonging to the EIF-3 subunit B family with an RRM (RNA recognition motif) domain. An important protein involved in protein translation/synthesis, eIF3B is known to bind to the 40S ribosomal subunit and promotes the binding of methionyl-tRNAi and mRNA. eIF3B is a component of the EIF3 multiprotein complex, composed of at least 13 different subunits. It is also known to interact with eIF3A and directly with the p170 subunit of EIF3, the region required for this association is located within the RRM. Ubiquitous expression of eIF3B is seen in most of the tissues. 
Specifications

Specifications

eIF3B
Polyclonal
Unconjugated
EIF3B
Liquid
EIF3B
Synthetic peptide (3 DAENVAVPEAAEERAEP 19) of eIF3B protein.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Immunohistochemistry (Paraffin), Western Blot
0.5 mg/mL
PBS with 0.05% BSA and 0.05% sodium azide
P55884, Q8JZQ9
EIF3-ETA, EIF3-P110, EIF3-P116, EIF3S9, PRT1, eIF-3-eta, eIF3 p110, eIF3 p116, eukaryotic translation initiation factor 3 subunit 9, eukaryotic translation initiation factor 3 subunit B, eukaryotic translation initiation factor 3, subunit 9 (eta, 116kD), eukaryotic translation initiation factor 3, subunit 9 eta, 116kDa, hPrt1, prt1 homolog
Rabbit
IgG
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
27979, 8662
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit