Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

eIF3j Rabbit anti-Canine, Equine, Human, Mouse, Non-human primate, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA523312

Catalog No. PIPA523312


Add to cart

Description

Description

eIF3J is an RNA-associated protein belonging to the eukaryotic translation initiation factor family. It belongs to the group of proteins involved in protein translation/synthesis where it binds to the 40S ribosomal subunit and promotes the binding of methionyl-tRNAi and mRNA to form the 40S initiation complex. eIF3J is a component of the eIF3 multiprotein complex, composed of at least 13 different subunits. It is also known to interact with eIF3S12. Ubiquitous expression is seen in most tissues.
Specifications

Specifications

eIF3J
Polyclonal
Unconjugated
EIF3J
Liquid
EIF3J, Eif3j1
Synthetic peptide (241 ADYGGYDGGYVQDYEDF 257) of eIF3J protein.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Canine, Equine, Human, Mouse, Non-human primate
Immunohistochemistry (Paraffin), Western Blot
0.5 mg/mL
PBS with 0.05% BSA and 0.05% sodium azide
O75822, Q3UGC7
PRO0391, EIF3S1, eIF3-alpha, eIF3-p35, eIF-3-alpha, eIF3 p35, eukaryotic translation initiation factor 3 subunit 1, eukaryotic translation initiation factor 3 subunit J, eukaryotic translation initiation factor 3, subunit 1 (alpha, 35kD), eukaryotic translation initiation factor 3, subunit 1 alpha, 35kDa
Rabbit
IgG
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
100070638, 487532, 78655, 8669
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit