Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

eukaryotic translation initiation factor 3, subunit J, Mouse, Polyclonal Antibody, Abnova™

Mouse polyclonal antibody raised against a full-length human EIF3S1 protein.

Manufacturer:  Abnova Corporation H00008669B01

Catalog No. 89-006-615


Add to cart

Description

Description

Eukaryotic initiation factor-3 (EIF3) has a molecular mass of about 600kDa and contains 13 nonidentical protein subunits, including EIF3J. EIF3 plays a central role in binding of initiator methionyl-tRNA and mRNA to the 40S ribosomal subunit to form the 40S initiation complex (Fraser et al., 2004 [PubMed 14688252]; Fraser et al., 2007 [PubMed 17588516]).[supplied by OMIM

Sequence: MAAAAAAAGDSDSWDADAFSVEDPVRKVGGGGTAGGDRWEGEDEDEDVKDNWDDDDDEKKEEAEVKPEVKISEKKKIAEKIKEKERQQKKRQEEIKKRLEEPEEPKVLTPEEQLADKLRLKKLQEESDLELAKETFGVNNAVYGIDAMNPSSRDDFTEFGKLLKDKITQYEKSLYYASFLEVLVRDVCISLEIDDLKKITNSLTVLCSEKQKQEKQSKAKKKKKGVVPGGGLKATMKDDLADYGGYDGGYVQDYEDFM
Specifications

Specifications

eukaryotic translation initiation factor 3, subunit J
Unconjugated
No additive
NM_003758.2
EIF3S1|eIF3-alpha|eIF3-p35
Mouse
50 uL
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Primary
8669
Western Blot
Mouse polyclonal antibody raised against a full-length human EIF3S1 protein.
EIF3J
Serum
EIF3J
EIF3S1 (NP_003749.2, 1 a.a. ~ 258 a.a) full-length human protein.
RUO
Y
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit