Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

eukaryotic translation initiation factor 3, subunit G, Mouse, Polyclonal Antibody, Abnova™

Mouse polyclonal antibody raised against a full-length human EIF3S4 protein.

Manufacturer:  Abnova Corporation H00008666B01

Catalog No. 89-006-614


Add to cart

Description

Description

Sequence: MPTGDFDSKPSWADQVEEEGEDDKCVTSELLKGIPLATGDTSPEPELLPGAPLPPPKEVINGNIKTVTEYKIDEDGKKFKIVRTFRIETRKASKAVARRKNWKKFGNSEFDPPGPNVATTTVSDDVSMTFITSKEDLNCQEEEDPMNKLKGQKIVSCRICKGDHWTTRCPYKDTLGPMQKELAEQLGLSTGEKEKLPGELEPVQATQNKTGKYVPPSLRDGASRRGESMQPNRRADDNATIRVTNLSEDTRETDLQELFRPFGSISRIYLAKDKTTGQSKGFAFISFHRREDAARAIAGVSGFGYDHLILNVEWAKPSTN
Specifications

Specifications

eukaryotic translation initiation factor 3, subunit G
Unconjugated
No additive
BC000733
EIF3-P42|EIF3S4|eIF3-delta|eIF3-p44
Mouse
50 uL
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Primary
8666
Western Blot
Mouse polyclonal antibody raised against a full-length human EIF3S4 protein.
EIF3G
Serum
EIF3G
EIF3S4 (AAH00733, 1 a.a. ~ 320 a.a) full-length human protein.
RUO
Y
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit