Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

eukaryotic translation initiation factor 3, subunit C, Mouse, Polyclonal Antibody, Abnova™

Mouse polyclonal antibody raised against a full-length human EIF3S8 protein.

Manufacturer:  Abnova Corporation H00008663B01

Catalog No. 89-006-611


Add to cart

Description

Description

Sequence: MSRFFTTGSDSESESSLSGEELVTKPVGGNYGKQPLLLSEDEEDTKRVVRSAKDKRFEELTNLIRTIRNAMKIRDVTKCLEEFELLGKAYGKAKSIVDKEGVPRFYIRILADLEDYLNELWEDKEGKKKMNKNNAKALSTLRQKIRKYNRDFESHITSYKQNPEQSADEDAEKNEEDSEGSSDEDEDEDGVSAATFLKKKSEAPSGESRKFLKKMDDEDEDSEDSEDDEDWDTGSTSSDSDSEEEEGKQTALASRFLKKAPTTDEDKKAAEKKREDKAKKKHDRKSKRLDEEEEDNEGGEWERVRGGVPLVKEKPKMFAKGTEITHAVVIKKLNEILQARGKKGTDRAAQIELLQLLVQIAAENNLGEGVIVKIKFNIIASLYDYNPNLATYMKPEMWGKCLDCINELMDILFANPNIFVGENILEESENLHNADQPLRVRGCILTLVERMDEEFTKIMQNTDPHSQEYVEHLKDEAQVCAIIERVQRYLEEKGTTEEVCRIYLLRILHTYYKFDYKAHQRQLTPPEGSSKSEQDQAENEGEDSAVLMERLCKYIYAKDRTDRIRTCAILCHIYHHALHSRWYQARDLMLMSHLQDNIQHADPPVQILYNRTMVQLGICAFRQGLTKDAHNALLDIQSSGRAKELLGQGLLLRSLQERNQEQEKVERRRQVPFHLHINLELLECVYLVSAMLLEIPYMAAHESDARRRMISKQFHHQLRVGERQPLLGPPESMREHVVAASKAMKMGDWKTCHSFIINEKMNGKVWDLFPEADKVRTMLVRKIQEESLRTYLFTYSSVYDSISMETLSDMFELDLPTVHSIISKMIINEELMASLDQPTQTVVMHRTEPTAQQNLALQLAEKLGSLVENNERVFDHKQGTYGGYFRDQKDGYRKNEGYMRRGGYRQQQSQTAY
Specifications

Specifications

eukaryotic translation initiation factor 3, subunit C
Unconjugated
No additive
NM_003752.3
EIF3S8|FLJ78287|MGC189744|eIF3-p110
Mouse
50 uL
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Primary
8663
Western Blot
Mouse polyclonal antibody raised against a full-length human EIF3S8 protein.
EIF3C
Serum
EIF3C
EIF3S8 (NP_003743.1, 1 a.a. ~ 913 a.a) full-length human protein.
RUO
Y
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit