Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

eIF4A2 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA527431

Catalog No. PIPA527431


Add to cart

Description

Description

PA5-27431 targets EIF4A2 in IF and WB applications and shows reactivity with Human samples. The PA5-27431 immunogen is recombit protein fragment corresponding to a region within amino acids 218 and 380 of Human EIF4A2. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

Specifications

Specifications

EIF4A2
Polyclonal
Unconjugated
EIF4A2
Liquid
EIF4A2
Recombit protein fragment corresponding to a region within amino acids 218 and 380 of Human EIF4A2
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
1 mg/mL
PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7
Q14240
BM-010, DDX2B, EIF4A, EIF4F, eIF-4A-II, eIF4A-II, ATP-dependent RNA helicase eIF4A-2, eukaryotic initiation factor 4A-II, eukaryotic translation initiation factor 4A
Rabbit
IgG
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
1974
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit