Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

eIF4A2 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA526198

Catalog No. PIPA526198


Add to cart

Description

Description

This antibody is predicted to react with mouse based on sequence homology.

Eukaryotic initiation factor 4A plays an important role in the binding of mRNA to the 43S preinitiation complex when protein synthesis begins. Two highly homologous forms of functional EIF4A genes, Eif4a1 and Eif4a2, have been isolated in mice; yeast cells also possess 2 EIF4A genes, TIF1 and TIF2. The murine Eif4a and yeast TIF genes appear to belong to a DEAD-box gene family, whose members exhibit extensive amino acid similarity and contain the asp-glu-ala-asp (DEAD) sequence. DEAD-box genes have been identified in species ranging from E-coli to humans. Their function appears to be related to transcriptional/translational regulation (referenced from OMIM).
Specifications

Specifications

EIF4A2
Polyclonal
PBS with 0.09% sodium azide
Q14240
BM-010, DDX2B, EIF4A, EIF4F, eIF-4A-II, eIF4A-II, ATP-dependent RNA helicase eIF4A-2, eukaryotic initiation factor 4A-II, eukaryotic translation initiation factor 4A
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
1974
Flow Cytometry, Immunohistochemistry, Western Blot
Unconjugated
EIF4A2
Liquid
EIF4A2
KLH conjugated synthetic peptide between 333-360 amino acids from the C-terminal region of human EIF4A2
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit