Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

eIF4B Rabbit anti-Human, Mouse, Non-human primate, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA523286

Catalog No. PIPA523286


Add to cart

Description

Description

eIF4B is a multidomain eukaryotic translation initiation factor with a range of activities. It contains one RRM (RNA recognition) domain, a consensus RNA-binding site (RNP-CS) near the N terminus, and a central domain rich in aspartic acid, arginine, tyrosine, and glycine (DRYG). The DRYG domain of eIF4B helps in self-association and interaction with the p170 subunit of eIF3. eIF4B binds to both mRNA and 18S ribosomal RNA and thus acts as a bridge between the mRNA and the 40S ribosomal subunit during translation initiation. The ATPase and RNA helicase activity of the DEAD box protein eIF4A is stimulated by eIF4B and thus functions in close association with eIF4F, a heterotrimeric protein that binds to mRNA cap structure and unwinds inhibitory secondary structure in the 5 ft. untranslated region of eucaryotic mRNAs in an ATP-dependent manner. Ubiquitous expression is seen in most tissues.
Specifications

Specifications

eIF4B
Polyclonal
Unconjugated
EIF4B
Liquid
EIF4B
Synthetic peptide (594 AALSVDGEDENEGEDYAE 611) of eIF4B protein.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Non-human primate
Western Blot
0.5 mg/mL
PBS with 0.05% BSA and 0.05% sodium azide
P23588, Q8BGD9
EIF-4B, PRO1843
Rabbit
IgG
100 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
1975, 75705
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit