Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ELF1 Goat anti-Human, Mouse, Polyclonal, Invitrogen™

Goat Polyclonal Antibody

Manufacturer:  Invitrogen PA131040

Catalog No. PIPA131040


Add to cart

Description

Description

PA1-31040 detects ELF1 from human and mouse samples. PA1-31040 has been successfully used in Western blot and ELISA applications. Positive controls of Jurkat or H460 lysates suggested. The PA1-31040 immunogen is synthetic peptide: AMKQNELLEPNSF, corresponding to amino acids 607-619 of Human ELF1. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

Specifications

Specifications

ELF1
Polyclonal
Unconjugated
ELF1
Liquid
Goat
IgG
100 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
13709, 1997
ELISA, Western Blot
0.5 mg/mL
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
P32519, Q60775
ELF1
synthetic peptide AMKQNELLEPNSF, corresponding to amino acids 607-619 of Human ELF1.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit