Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ELF3 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA14221

Catalog No. PA14221


Add to cart

Description

Description

PA1-4221 detects the ESE-1 protein in human samples. PA1-4221 has been successfully used in Western blot procedures. By Western blot this antibody detects both isoforms of ESE-1 protein, both bands around ~40 kDa, in T84 colon carcinoma cells. The PA1-4221 immunogen is a synthetic peptides corresponding to residues M(1) A A T C E I S N I F S N Y F S(16) and S(335) S G W K E E E V L Q S R N(348) of the N- and C-termini of human ESE-1.

ESE-1 (for epithelium-specific Ets), is a novel, highly tissue-restricted member of the ets transcription factor/oncogene family, which has features distinct from those of any other ets-related factor. ESE-1 binds with high affinity to and transactivates the ets binding site in the promoter of the keratinocyte terminal differentiation marker gene, SPRR2A. ESE-1 is a critical regulator of epithelial cell differentiation.
Specifications

Specifications

ELF3
Polyclonal
Unconjugated
ELF3
Liquid
ELF3
Synthetic peptides corresponding to residues M(1) A A T C E I S N I F S N Y F S(16) and S(335) S G W K E E E V L Q S R N(348) of human ESE-1.
Affinity chromatography
RUO
Antibody
Polyclonal
Human
Western Blot
1 mg/mL
PBS with 0.02% sodium azide
P78545
Epithelium-Specific Ets
Rabbit
IgG
100 μg
Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C
Primary
1999
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit