Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ELF3 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA526451

Catalog No. PIPA526451

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

ESE-1 (Epithelium-Specific Ets) is a novel, highly tissue-restricted member of the ets transcription factor/oncogene family, which has features distinct from those of any other ets-related factor. ESE-1 binds with high affinity to and transactivates the ets binding site in the promoter of the keratinocyte terminal differentiation marker gene, SPRR2A. ESE-1 is a critical regulator of epithelial cell differentiation.
Specifications

Specifications

ELF3
Polyclonal
PBS with 0.09% sodium azide
P78545
EPR-1, ERT, ESE-1, ESX, E74-like factor 3 (ETS domain transcription factor, serine box, epithelial-specific), E74-like factor 3 (ets domain transcription factor), ETS-related transcription factor Elf-3, epithelial-restricted with serine box, epithelium-restricted Ets protein ESX, epi
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
1999
Western Blot
Unconjugated
ELF3
Liquid
ELF3
KLH conjugated synthetic peptide between 322-349 amino acids from the C-terminal region of human ELF3
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit