Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ELL Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA523680
Description
Specifications
ELL | |
Polyclonal | |
ELL | |
C19orf17, ELL1, MEN, PPP1R68, ELL gene (11-19 lysine-rich leukemia gene), RNA polymerase II elongation factor ELL, eleven-nineteen lysine-rich leukemia protein, protein phosphatase 1, regulatory subunit 68 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
13716, 8178 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
Unconjugated | |
P55199, O08856 | |
ELL | |
KLH conjugated synthetic peptide between 516-545 amino acids from the C-terminal region of human ELL | |
400 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction