Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ELL Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA526488

Catalog No. PIPA526488

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Elongation factor that can increase the catalytic rate of RNA polymerase II transcription by suppressing transient pausing by the polymerase at multiple sites along the DNA.
Specifications

Specifications

ELL
Polyclonal
PBS with 0.09% sodium azide
P55199, O08856
C19orf17, ELL1, MEN, PPP1R68, ELL gene (11-19 lysine-rich leukemia gene), RNA polymerase II elongation factor ELL, eleven-nineteen lysine-rich leukemia protein, protein phosphatase 1, regulatory subunit 68
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
13716, 8178
Western Blot
Unconjugated
ELL
Liquid
ELL
KLH conjugated synthetic peptide between 516-545 amino acids from the C-terminal region of human ELL
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit