Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
elongation factor RNA polymerase II-like 3 (B01), Mouse anti-Human, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a full-length human ELL3 protein.
Supplier: Abnova Corporation H00080237B01
Description
Sequence: MEELQEPLRGQLRLCFTQAARTSLLLLRLNDAALRALQECQRQQVRPVIAFQGHRGYLRLPGPGWSCLFSFIVSQCCQEGAGGSLDLVCQRFLRSGPNSLHCLGSLRERLIIWAAMDSIPAPSSVQGHNLTEDARHPESWQNTGGYSEGDAVSQPQMALEEVSVSDPLASNQGQSLPGSSREHMAQWEVRSQTHVPNREPVQALPSSASRKRLDKKRSVPVATVELEEKRFRTLPLVPSPLQGLTNQDLQEGEDWEQEDEDMGPRLEHSSSVQEDSESPSPEDIPDYLLQYRAIHSAEQQHAYEQDFETDYAEYRILHARVGTASQRFIELGAEIKRVRRGTPEYKVLEDKIIQEYKKFRKQYPSYREEKRRCEYLHQKLSHIKGLILEFEEKNRGSSpecifications
elongation factor RNA polymerase II-like 3 | |
Polyclonal | |
Mouse polyclonal antibody raised against a full-length human ELL3 protein. | |
ELL3 | |
FLJ22637 | |
Mouse | |
50 μL | |
Transcription Regulation | |
Primary | |
Human | |
Serum |
Western Blot | |
Unconjugated | |
No additive | |
BC019293 | |
ELL3 | |
ELL3 (AAH19293, 1 a.a. ∼ 397 a.a) full-length human protein. | |
RUO | |
Yes | |
80237 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title