Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ELOVL4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA131206

Catalog No. PA131206


Add to cart

Description

Description

PA1-31206 detects ELOVL4 from mouse, rat, human samples. PA1-31206 has been successfully used in Western blot applications. The PA1-31206 immunogen is: Synthetic peptide: SYICQSVDYSNNVHE, corresponding to Internal sequence amino acids 107-121 of Human ELOVL4. Store at 4°C short term. For extended storage aliquot and store at -20°C or below. Avoid freeze-thaw cycles.

This gene encodes a membrane-bound protein which is a member of the ELO family, proteins which participate in the biosynthesis of fatty acids. Consistent with the expression of the encoded protein in photoreceptor cells of the retina, mutations and small deletions in this gene are associated with Stargardt-like macular dystrophy (STGD3) and autosomal domit Stargardt-like macular dystrophy (ADMD), also referred to as autosomal domit atrophic macular degeneration.
Specifications

Specifications

ELOVL4
Polyclonal
Unconjugated
ELOVL4
Liquid
ELOVL4
synthetic peptide SYICQSVDYSNNVHE, corresponding to Internal sequence amino acids 107-121 of Human ELOVL4.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Western Blot
0.68 mg/mL
Antibody with no preservative
Q9GZR5, Q9EQC4
ADMD, FLJ17667, FLJ92876, STGD2, STGD3
Rabbit
IgG
100 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
315851, 6785, 83603
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit