Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.

anti-ELOVL7, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP191349

Catalog No. NBP191349



Description

Description

ELOVL7 Polyclonal antibody specifically detects ELOVL7 in Human samples. It is validated for Western Blot.
Specifications

Specifications

ELOVL7
Unconjugated
PBS and 2% Sucrose with 0.09% Sodium Azide
Purified
ELOVL7
Synthetic peptide directed towards the N terminal of human ELOVL7. Peptide sequence MAFSDLTSRTVHLYDNWIKDADPRVEDWLLMSSPLPQTILLGFYVYFVTS.
Protein A purified
RUO
Primary
79993
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 100μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer.
Western Blot
Western Blot 1:1000
NP_079206
ELOVL family member 7, elongation of long chain fatty acids, ELOVL fatty acid elongase 7
Rabbit
IgG
100ul
Store at -20C. Avoid freeze-thaw cycles.
Polyclonal
Expected identity based on immunogen sequence: Human: 100%; Canine: 85%; Equine: 85%; Pig: 85%; Rat: 85%.
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only