Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

emerin, Mouse, Polyclonal Antibody, Abnova™

Mouse polyclonal antibody raised against a full-length human EMD protein.

Manufacturer:  Abnova Corporation H00002010B01

Catalog No. 89-005-298


Add to cart

Description

Description

Emerin is a serine-rich nuclear membrane protein and a member of the nuclear lamina-associated protein family. It mediates membrane anchorage to the cytoskeleton. Dreifuss-Emery muscular dystrophy is an X-linked inherited degenerative myopathy resulting from mutation in the emerin gene. [provided by RefSeq

Sequence: MDNYADLSDTELTTLLRRYNIPHGPVVGSTRRLYEKKIFEYETQRRRLSPPSSSAASSYSFSDLNSTRGDADMYDLPKKEDALLYQSKGYNDDYYEESYFTTRTYGEPESAGPSRAVRQSVTSFPDADAFHHQVHDDDLLSSSEEECKDRERPMYGRDSAYQSITHYRPVSASRSSLDLSYYPTSSSTSFMSSSSSSSSWLTRRAIRPENRAPGAGLGQDRQVPLWGQLLLFLVFVIVLFFIYHFMQAEEGNPF
Specifications

Specifications

emerin
Unconjugated
No additive
BC000738
EDMD|LEMD5|STA
Mouse
50 uL
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Primary
2010
Immunofluorescence, Western Blot
Mouse polyclonal antibody raised against a full-length human EMD protein.
EMD
Serum
EMD
EMD (AAH00738, 1 a.a. ~ 254 a.a) full-length human protein.
RUO
Y
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit