Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-EML1, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP130956

Catalog No. NBP130956


Add to cart

Description

Description

EML1 Polyclonal antibody specifically detects EML1 in Human, Mouse, Rat, Monkey samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications

Specifications

EML1
1.0mg/mL
Western Blot 1:500-1:3000, Immunohistochemistry 10 - 1:500, Immunocytochemistry/Immunofluorescence 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000
Affinity Purified
EML1
Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human EML1. The exact sequence is proprietary.
Antigen Affinity-purified
RUO
Primary
2009
Human, Monkey, Mouse, Rat
Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
PBS (pH 7.0), 1.0% BSA and 20% Glycerol with 0.01% Thimerosal
echinoderm microtubule associated protein like 1, ELP79, EMAP1, EMAP-1, EMAPechinoderm microtubule-associated protein-like 1, EMAPL1, EMAPLhuEMAP-1, FLJ45033, HuEMAP, HuEMAP-1
Rabbit
IgG
100 ul
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Polyclonal
Tested in Human, Mouse, Monkey, and Rat. Predicted cross-reactivity based on sequence identity: Zebrafish (85%). Not yet tested in any other species.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only